Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556959 Supplier Invitrogen™ Supplier No. PA556959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASPAQYTTRI SPVCLASSNE ALTEGLTCVT TGWGRLSGVG NVTPAHLQQV ALPLVTVNQC RQYWGSSITD SMICAGGAGA SSCQG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 88%.

The CTRL gene encodes chymotrypsin-like protein, a serine protease involved in the digestion of proteins by cleaving peptide bonds. It belongs to the chymotrypsin family of enzymes that are predominantly expressed in the pancreas and play crucial roles in the breakdown of dietary proteins in the small intestine. CTRL functions by hydrolyzing specific peptide bonds, contributing to efficient protein digestion and nutrient absorption. This enzyme's activity is tightly regulated to prevent premature activation within the pancreas, which could lead to pancreatitis or other digestive disorders. The study of the CTRL gene is significant in understanding the mechanisms of digestive health and in diagnosing pancreatic disorders. Furthermore, CTRL's enzyme activity and its role in proteolytic pathways make it an important target for research in digestive enzyme supplementation therapies and treatment strategies for conditions involving impaired protein digestion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ctrl
Gene Accession No. P40313
Gene Alias 0910001G08Rik; 1810004D15Rik; AV005227; chymopasin; chymotrypsin A CTRA-1; chymotrypsin like; chymotrypsin-like; chymotrypsin-like protease CTRL-1; Ctra1; Ctra-1; CTRL; CTRL1; mFLJ00366
Gene Symbols Ctrl
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CTRL. Recombinant protein control fragment (Product #RP-95843).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1506
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.