Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTRP1 Monoclonal Antibody (C2), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA550947
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550947 200 μL
FEMA550948 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA550947 Supplier Invitrogen™ Supplier No. MA550947
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. In obese (ob/ob) mice, RT-PCR analysis showed that mCTRP1 transcripts are seen at substantially higher levels in adipose tissues compared to those of normal mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene C1QTNF1
Gene Accession No. Q9BXJ1
Gene Alias 1600017K21Rik; C1q and tumor necrosis factor related protein 1; C1qtnf1; Complement C1q tumor necrosis factor-related protein 1; Ctrp1; G protein coupled receptor interacting protein; g protein-coupled receptor-interacting protein; GIP; putative secreted protein ZSIG37; UNQ310/PRO353; Zsig37
Gene Symbols C1QTNF1
Host Species Mouse
Immunogen Recombinant Human C1QTNF1 protein, Arg26-Pro281 (Accession #Q9BXJ1).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100217377, 114897
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.