Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584486 Supplier Invitrogen™ Supplier No. PA584486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAEESRDAEP RRELLCSGRP WTWRAAARVT TMIPWVLLAC ALPCAADPLL GASARRDFRK GSPQLVCSLP G Highest antigen sequence indentity to the following orthologs: Mouse - 61%, Rat - 52%.

CTRP2, also known as adiponectin C1q and collagen domain-containing protein 2, is a protein that belongs to the C1q/TNF-related protein family. It is expressed in various tissues, including adipose tissue, skeletal muscle, and the heart, and is involved in the regulation of energy metabolism and insulin sensitivity. CTRP2 has been shown to have anti-inflammatory and cardioprotective effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1QTNF2
Gene Accession No. Q9BXJ5
Gene Alias 1810033K05Rik; C1q and tumor necrosis factor related protein 2; C1qtnf2; Complement C1q tumor necrosis factor-related protein 2; complement-c1q tumor necrosis factor-related protein 2; Crtp2; CTRP2; mCTRP2; RGD1561041; UNQ6349/PRO21054; zacrp2
Gene Symbols C1QTNF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1QTNF2. Recombinant protein control fragment (Product #RP-101345).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114898
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.