Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114920 Supplier Invitrogen™ Supplier No. PA5114920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C1QTNF2.

CTRP2, also known as adiponectin C1q and collagen domain-containing protein 2, is a protein that belongs to the C1q/TNF-related protein family. It is expressed in various tissues, including adipose tissue, skeletal muscle, and the heart, and is involved in the regulation of energy metabolism and insulin sensitivity. CTRP2 has been shown to have anti-inflammatory and cardioprotective effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene C1QTNF2
Gene Accession No. Q9BXJ5
Gene Alias 1810033K05Rik; C1q and tumor necrosis factor related protein 2; C1qtnf2; Complement C1q tumor necrosis factor-related protein 2; complement-c1q tumor necrosis factor-related protein 2; Crtp2; CTRP2; mCTRP2; RGD1561041; UNQ6349/PRO21054; zacrp2
Gene Symbols C1QTNF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human C1QTNF2(Accession Q9BXJ5), corresponding to amino acid residues K96-G146.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114898, 497886
Target Species Human, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.