Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTRP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564194
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564194 Supplier Invitrogen™ Supplier No. PA564194
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MYSYEMKGKS DTSSNHAVLK LAKGDEVWLR MGNGALHGDH QRFSTFAGFL LFET Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of human CTRP3 have been reported. It has been suggested that CTRP3 may play a role in skeletal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1qtnf3
Gene Accession No. Q9BXJ4
Gene Alias 2310005P21Rik; C1ATNF3; C1q and tumor necrosis factor related protein 3; C1qtnf3; cartducin; cartonectin; collagenous repeat-containing sequence 26 kDa protein; collagenous repeat-containing sequence of 26-kDa; complement C1q tumor necrosis factor-related protein 3; Corcs; CORS; CORS26; CORS-26; CTRP3; Secretory protein CORS26; UNQ753/PRO1484
Gene Symbols C1qtnf3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CTRP3. Recombinant protein control fragment (Product #RP-103925).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.