Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTRP3 Polyclonal Antibody
CTRP3 Polyclonal Antibody
Supplier: Thermo Scientific PA584704
Description
CTRP3 Polyclonal Antibody for ICC/IF

Specifications
CTRP3 | |
Polyclonal | |
Unconjugated | |
C1qtnf3 | |
2310005P21Rik; C1ATNF3; C1q and tumor necrosis factor related protein 3; C1qtnf3; cartducin; cartonectin; collagenous repeat-containing sequence 26 kDa protein; collagenous repeat-containing sequence of 26-kDa; complement C1q tumor necrosis factor-related protein 3; Corcs; CORS; CORS26; CORS-26; CTRP3; Secretory protein CORS26; UNQ753/PRO1484 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114899 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BXJ4 | |
C1qtnf3 | |
Recombinant protein corresponding to Human C1QTNF3. Recombinant protein control fragment (Product #RP-107472). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction