Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTRP6 Polyclonal Antibody

CTRP6 Polyclonal Antibody
Supplier: Thermo Scientific PA520155
Description
CTRP6 Polyclonal Antibody for Western Blot, IHC
Specifications
| CTRP6 | |
| Polyclonal | |
| Unconjugated | |
| C1QTNF6 | |
| 2810036M19Rik; C1q and tumor necrosis factor related protein 6; C1qtnf6; Complement C1q tumor necrosis factor-related protein 6; complement-c1q tumor necrosis factor-related protein 6; CTFP6; CTRP6; UNQ581/PRO1151; ZACRP6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114904, 315114, 72709 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q3T1I2, Q6IR41, Q9BXI9 | |
| C1QTNF6 | |
| A 15 amino acid peptide from near the center of human CTRP6. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction