Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTRP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551674 Supplier Invitrogen™ Supplier No. PA551674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQRCCDSEDP LDPAHVSSAS SSGRPHALPE IRPYINITIL KGDKGDPGPM GLPGYMGREG PQGEPGPQGS KGDKGEMGSP GAPCQKRFFA FSVGRKTALH SGEDFQTLLF ERVFVNLDGC FDMATGQFAA PLR Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 66%.

Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. CTRP6 contains at least 4 glycosylation motifs, suggesting that CTRP6 may be highly post-translationally modified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRP6
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1QTNF6
Gene Accession No. Q9BXI9
Gene Alias 2810036M19Rik; C1q and tumor necrosis factor related protein 6; C1qtnf6; Complement C1q tumor necrosis factor-related protein 6; complement-c1q tumor necrosis factor-related protein 6; CTFP6; CTRP6; UNQ581/PRO1151; ZACRP6
Gene Symbols C1QTNF6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CTRP6. Recombinant protein control fragment (Product #RP-101999).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114904
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.