Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTRP6 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177218SS
Description
CTRP6 Polyclonal specifically detects CTRP6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CTRP6 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
C1q and tumor necrosis factor related protein 6, complement C1q tumor necrosis factor-related protein 6, complement-c1q tumor necrosis factor-related protein 6, CTRP6ZACRP6 | |
Rabbit | |
Affinity Purified | |
Primary | |
These proteins often migrate in SDS-PAGE at positions other than their predicted size. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL | |
AAQ88740 | |
C1QTNF6 | |
Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human CTRP6. The immunogen is located within the last 50 amino acids of CTRP6. Amino Acid Squence: RLFKRQRENAIYSND | |
0.025 mg | |
114904 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction