Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ OpTmizer™ One SFM

Catalog No. p-200019995
Click to view available options
Quantity:
1 L
5 L
10 L
Format:
BPC
Bottle

Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies.

Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies. This medium improves cell count and early memory phenotype throughout all stages of expansion.

Benefits:

  • Provides enhanced T-cell expansion and expression of early memory phenotype makers compared to other chemically defined media
  • Demonstrates flexibility across culture vessels. In both G-Rex™ plates and rocking bioreactors, it supports scale-up while maintaining desired T-cell phenotype.
  • Expands both healthy and disease state donors
  • Supports efficient viral and non-viral editing with downstream expansion
  • Mitigates risk of contaminants and variability associated with animal-sourced materials
  • Supports scale-up via larger catalog pack sizes

Features:

  • Animal-origin-free (AOF) formulation
  • Serum-free—designed without the need for serum or serum replacement
  • No separate components—no need to mix basal medium with supplement to obtain complete medium
  • Yields high density cultures
  • Supports T-cell activation using CTS Dynabeads (both passive release CD3/CD28 (Cat. No.40203D) and detachable CD3/CD28 (Cat No. A56992))
  • Available in 1 L bottle, 1 L bag, 5 L bag, and 10 L bag options

Increased efficiency and consistent performance:

CTS OpTmizer One SFM is an AOF formulation that supports high density cultures in static and dynamic vessels. It displays consistent performance for T-cell expansion, with better early memory phenotype at early stages.

Simplify your transition into clinical research:

The Gibco Cell Therapy Systems (CTS) portfolio offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. CTS products deliver:

  • Reduced burden in qualifying reagents
  • Appropriate certificates of analysis and origin available
  • Convenient access to our Drug Master File (DMF)

Specifications

Classification Animal Origin-free, Serum-free, cGMP
Form Liquid
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives None
Without Additives No Glutamine, No Phenol Red
Product Line CTS™ OpTmizer™
Quantity 1 L
Format Bottle
Content And Storage Store at 2-8°C and protect from light.
Culture Type Suspension Cell Culture, Dynamic Bioreactors
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.