Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ Rotea™ Single-Use Kit

Catalog No. p-7203095
Encompass
Click to view available options
Quantity:
10 Kits

The Gibco CTS Rotea Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System. The kit is capable of flow rates of 5–110 mL/min (see Hi-Flow kit for higher rates).

The Gibco CTS Rotea Single-Use Kit can perform a wide range of cell processing steps, making it ideal for wash & concentrate, separation, isolation, and more. The single-use kit provides flexibility for process development and easily supports commercial-scale manufacturing. Each kit is compatible with sterile luer connectors or thermal welding.

Tubing material: DEHP-free PVC

Tube dimensions: 0.160 in. OD, 0.116 in. ID

Pump tube material: Bioprene™ plastic

Kits are gamma-irradiated and individually packaged.

TRUSTED_SUSTAINABILITY

Specifications

Material Tritan MX711 Copolyester (Chamber), DEHP-free PVC (Tubing)
For Use With (Equipment) CTS Rotea Counterflow Centrifugation System
Disposable Single-use
Capacity 20 L
Max. RCF 3000 x g
Certifications/Compliance ISO 13485
Quantity 10 Kits
Sterilization Method Gamma-irradiated
Cell Type Agnostic
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Flow Rate 160 mL/min.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.