Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ StemPro™ MSC SFM

Catalog No. a1033201
Encompass_Preferred
Click to view available options
With Additives:
Low Glucose, Sodium Pyruvate, Sodium Bicarbonate, Adenosine, Deoxyribonucleosides
Without Additives:
No Glutamine, No Phenol Red
This item is not returnable. View return policy
This item is not returnable. View return policy

StemPro™ MSC SFM is a serum-free medium (SFM) specially formulated for the growth and expansion of human mesenchymal stem cells (MSCs).

StemPro™ MSC SFM is a serum-free medium (SFM) specially formulated for the growth and expansion of human mesenchymal stem cells (MSCs). StemPro™ MSC SFM enables superior human MSC growth and increased consistency compared to classical serum-supplemented medium (DMEM + 10% FBS).

Using StemPro™ MSC SFM, human MSCs can be expanded beyond 5 passages while still maintaining their tri-lineage mesoderm differentiation potential (i.e., ability to differentiate into osteogenic, chondrogenic and adipogenic lineages). Each lot of StemPro™ MSC SFM is qualified using a human MSC performance assay and human MSCs growing in serum-supplemented medium can be transitioned directly into StemPro™ MSC SFM with little or no adaptation required. In addition, StemPro™ MSC SFM facilitates expansion of MSCs directly from primary human bone marrow.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mesenchymal Stem Cells
Classification Serum-free
Form Liquid
Product Type Stem Cell Serum Free Media (SFM)
Serum Level Serum-free
With Additives Low Glucose, Sodium Pyruvate, Sodium Bicarbonate, Adenosine, Deoxyribonucleosides
Without Additives No Glutamine, No Phenol Red
Product Line StemPro™
Quantity 1 Kit
Endotoxin Level Low (≤10 EU/mL)
Inorganic Salts Calcium, Sodium Phosphate
Purity or Quality Grade Research Grade
Species Human
Content And Storage This kit is shipped in two parts with separate storage requirements. These components are not sold individually:
• StemPro™ MSC SFM Basal Medium, 500 mL (ships at room temperature, store in the dark at 2°C to 8°C)
• StemPro™ MSC SFM Supplement, 75 mL (ships on dry ice, store in the dark at -5°C to -20°C)
Culture Environment CO2
Culture Type Adherent Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.