Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ TrueCut™ Cas9 Protein

Catalog No. p-9681002
Encompass_Preferred
Click to view available options
Quantity:
2.5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Gibco™ CTS TrueCut Cas9 Protein is our highest performing CRISPR-Cas9 protein, engineered to deliver maximum ribonucleoprotein (RNP)-editing efficiency.

Gibco CTS TrueCut Cas9 Protein is our highest performing CRISPR-Cas9 protein, engineered to deliver maximum ribonucleoprotein (RNP)-editing efficiency. This protein is manufactured in compliance with standards for Ancillary Materials for Cell, Gene and Tissue-Based Products and is subjected to stringent quality specifications. It is provided in a high-concentration and large-scale format for process development and clinical research applications.

Features of CTS TrueCut Cas9 Protein include:
Consistent high editing efficiency—superior CRISPR-Cas9 editing efficiency in all tested cell lines including primary immune cells (>90% editing in primary T cells)
Higher stringency manufacturing—manufactured to meet the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products including USP <1043>, Ph.Eur. 5.2.12, and ISO 20399, following the principles of 21 CFR Part 820 in a FDA-registered manufacturing site
Aseptic manufacturing—entire process completed using single-use technology
Extensive safety testing—includes sterility, endotoxin, mycoplasma, residual nucleic acids, and proteins
Sterile filling—finished product is vialed in a Class A/B (Class 100 and ISO 5 equivalents) cleanroom
Traceability documentation—COA, COO, Drug Master Files, and Regulatory Support Files are available

CTS TrueCut Cas9 Protein is a Streptococcus pyogenes Cas9 (wt) protein, produced by recombinant DNA technology in E. coli for genome editing with CRISPR technology. Cas9 protein forms a very stable RNP complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. Incorporation of NLS aids delivery to the nucleus, increasing the editing efficiency and on-target cleavage.

If you would like to evaluate our TrueCut Cas9 protein or conduct small-scale research experiments, we recommend using our TrueCut Cas9 Protein v2 (Cat. No. A36497).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 mg/mL
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Quantity 2.5 mg
Product Type Cas9 Protein
Sterility Sterile-filtered
Content And Storage Store at –20°C.
Shipping Condition Dry Ice
Expression System E. coli
Form Liquid
Research Category Genome Editing
Manufacturing Quality ISO13485
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use or Non-Commercial Manufacturing of Cell-Based Products for Clinical Research. Caution: Not intended for direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.