Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ Viral Production Cells

Catalog No. A53956
Encompass_Preferred
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53956 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A53956 Supplier Gibco™ Supplier No. A53956
Add to Cart
Edge
Add to Cart
Request a Quote

CTS Viral Production Cells are derived from the human HEK293 cell line and are a core component of the CTS LV-MAX lentiviral production system to produce maximal lentiviral titers.

CTS Viral Production Cells are derived from the human HEK293 cell line and are a core component of the CTS LV-MAX lentiviral production system to produce maximal lentiviral titers. The cells allow high density, serum-free suspension culture with minimal cell clumping in CTS LV-MAX Production Medium. They are specifically validated for lentiviral vector production for the purpose of cell and gene therapy applications, but may be suitable for the production of other viral vectors where the absence of SV40 large T antigen in HEK293 cells is desired. These cGMP-banked cells have gone through a documented validation process to ensure that they can be transfected with multiple plasmids and that viable viral particles can be produced.

Features of the CTS Viral Production Cells include:

  • Fully-documented cGMP-banked cell line (HEK293F-derived) with detailed cell line lineage history
  • Extensive quality testing based on the US FDA and European Medicines Agency (EMA) guidances for therapeutic use
  • Flexible licensing options without royalties

Please visit www.thermofisher.com/celllicenserequest for more information and access.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 107 cells in a 2-mL vial. Store in vapor phase liquid nitrogen.
Cell Line 293F
Cell Type Kidney (Embryonic)
Culture Type Suspension Cell Culture
Format Cryopreserved vial
For Use With (Application) Cell & Gene Therapy Research, Development, and Manufacturing
Media Recommendation Gibco™ CTS™ Viral Production Medium
No. of Cells See Certificate of Analysis
Product Type Viral Production Cell Line
Quantity 1.5 mL
Shipping Condition Dry ice or vapor phase liquid nitrogen
Tested For LV Production
Manufacturing Quality 21 CFR Parts 210 and 211
Show More Show Less
Are CTS Viral Production Cells 293T-derived?

No, CTS Viral Production Cells are HEK 293F suspension cells that have been adapted to CTS LV-MAX Production Medium. They do not express the SV40 large T antigen.

How do I bank CTS Viral Production Cells?

CTS Viral Production Cells can be frozen directly in cryogenic medium consisting of 90% CTS LV-MAX Production Medium plus 10% DMSO. Here is the protocol:

- Bank cells at passage 3 when cell density reaches ˜3.0-4.5 x 10E6 viable cells/mL at >95% viability.
- Centrifuge the cells at 100 x g (low speed) for 5 mins to pellet. Discard the spent medium.
- Prepare freezing medium: e.g., 10 mL of freezing medium is 9 mL of CTS LV-MAX Production Medium + 1 mL DMSO; mix well and sterilize by 0.2 µm filtration.
- Resuspend the cell pellet in the prepared freezing medium.
- Dilute the cells to a final density of 10 x10E6 viable cells/mL and aliquot 1 mL per cryovial.
- Freeze the cells at -80 degrees C for 1 day.
- Transfer frozen vials to liquid nitrogen for long-term storage.

Can I use CTS Viral Production Cells (Cat. No. A53956) with CTS AAV MAX products?

CTS Viral Production Cells 2.0 are specifically designed to work with CTS AAV MAX products to provide maximal adeno-associated virus (AAV) titers and optimal performance. CTS Viral Production Cells can be used but we don't recommend doing this as it will reduce system performance.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.