Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ Vitronectin (VTN-N) Recombinant Human Protein, Truncated

Catalog No. A27940
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27940 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27940 Supplier Gibco™ Supplier No. A27940

CTS™ (Cell Therapy Systems) Vitronectin (VTN-N) Recombinant Human Protein provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs).

  • Provides a defined surface for feeder-free culture of human pluripotent stem cells (PSCs)
  • When used with Essential 8™ Medium (Cat. No. A1517001), CTS™ Vitronectin has been proven to maintain pluripotency and normal growth characteristics in multiple PSC lines
  • Shown to support PSC growth for >8 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages
  • VTN-N variant of vitronectin supports human pluripotent stem cell attachment and survival better than wild-type vitronectin when used in conjunction with Essential 8 Medium (1)
  • Designed for clinical research—USP Chapter 1043 considered in the design
  • Animal origin–free—for reduced variability and contamination risk in PSC cultures
  • Optimized—designed for use with Essential 8 Medium
  • Consistent—supports seamless transition from Vitronectin
  • Traceable—documentation includes COO, COA, and DMF
  • Controlled—manufactured in compliance with ISO-13485:2003

Designed for clinical research

  • USP Chapter 1043 has been considered in the design of CTS™ Vitronectin, and this product is the only such product labeled with 'For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration to humans or animals.'
  • Harmonized documentation is provided with the product and includes a Certificate of Analysis (COA), Certificate of Origin (COO), and Drug Master File (DMF) to support regulatory filings.

Animal origin–free system

  • CTS™ Vitronectin is animal origin–free at the primary level to help reduce contamination risk and variability associated with animal-origin raw materials compared to human plasma–derived vitronectin and standard basement membrane extraCTS™ (BMEs)

Optimized and defined surface

  • Unlike standard basement membrane extraCTS™ (BMEs), CTS™ Vitronectin is based on the N-variant design of vitronectin (VTN-N) from the laboratory of James Thomson for use with Essential 8 Medium (1) and has been shown to support human pluripotent stem cell attachment and survival better than wild type vitronectin
  • Provides an optimal environment for feeder-free pluripotent stem cell culture.

Seamless transition from bench to clinic

  • PSC cultures expanded in other feeder or feeder-free systems can be transitioned to Essential 8 Medium and CTS™ Vitronectin with minimal adaptation

Manufacturing control and traceability

  • CTS™ Vitronectin is manufactured at a site that is FDA registered and has an ISO13485-certified quality management system
  • Full traceability documentation such as Certificate of Origin, Certificate of Analysis, and Drug Master File are available
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mL
Species Human
Recombinant Recombinant
Content And Storage Supplied at a concentration of 0.9 mg/mL.
Store at -80°C and protect from light.
Classification Animal Origin-Free
Shipping Condition Dry Ice
Protein Form Recombinant
Form Liquid
Protein Subtype Vitronectin
Product Line CTS

Designed for clinical researchUSP Chapter 1043 has been considered in the design of CTS™ Vitronectin, and this product is the only such product labeled with For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration to humans or animals. Harmonized documentation is provided with the product and includes a Certificate of Analysis (COA) and Certificate of Origin (COO) to support regulatory filings.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.