Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTSD Mouse anti-Human, Unconjugated, Clone: CTSD/2781, Abnova™

Catalog No. 89337398 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-398 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-398 Supplier Abnova Corporation Supplier No. MAB20903
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CTSD Monoclonal antibody specifically detects CTSD in Human samples. It is validated for IHC-P.

Specifications

Antigen CTSD
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CTSD/2781
Conjugate Unconjugated
Description cathepsin D
Gene CTSD
Gene Alias CLN10, CPSD, MGC2311
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 104-250 of human CTSD
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1509
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.