Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTSK Mouse anti-Human, Unconjugated, Clone: CTSK/2791, Abnova™

Catalog No. 89337659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-659 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-659 Supplier Abnova Corporation Supplier No. MAB21166
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CTSK Monoclonal antibody specifically detects CTSK in Human samples. It is validated for IHC-P.

Specifications

Antigen CTSK
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CTSK/2791
Conjugate Unconjugated
Description cathepsin K
Gene CTSK
Gene Alias CTS02, CTSO, CTSO1, CTSO2, MGC23107, PKND, PYCD
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 163-274 of human CTSK
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1513
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.