Learn More
CTSL1 Mouse anti-Human, Clone: OTI5C8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812846

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. At least two transcript variants encoding the same protein have been found for this gene.Specifications
CTSL1 | |
Monoclonal | |
Unconjugated | |
CTSL | |
CATL, CTSL1, MEP | |
Mouse | |
Affinity Chromatography | |
RUO | |
1514 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI5C8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P07711 | |
CTSL | |
Human recombit protein fragment corresponding to amino acids 114-333 of human CTSL produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.