Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTTNBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100461 Supplier Invitrogen™ Supplier No. PA5100461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CTTNBP2.

CTTB2 is a protein with six ankyrin repeats and several proline-rich regions. A similar gene in rat interacts with a central regulator of the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTTNBP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTTNBP2
Gene Accession No. B9EJA2, Q8WZ74
Gene Alias 3010022N24Rik; 4732477G22Rik; 6430526E05; 9130022E09Rik; AU040881; brain specific cortactin-binding protein CBP90; C7orf8; Cbp90; cortactin binding protein 2; cortactin-binding protein 2; cortactin-binding protein 2 intron retention form; CORTBP2; Cttnbp2; CTTNBP2 intron retention form; KIAA1758; mKIAA1758; ORF4
Gene Symbols CTTNBP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTTNBP2(Accession Q8WZ74), corresponding to amino acid residues N1475-D1525.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30785, 83992
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.