Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTTNBP2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS14107R

Description
CTTB2 is a protein with six ankyrin repeats and several proline-rich regions. A similar gene in rat interacts with a central regulator of the actin cytoskeleton.Specifications
CTTNBP2 | |
Polyclonal | |
Unconjugated | |
CTTNBP2 | |
3010022N24Rik; 4732477G22Rik; 6430526E05; 9130022E09Rik; AU040881; C7orf8; cortactin binding protein 2; Cortactin-binding protein 2; cortactin-binding protein 2 intron retention form; CORTBP2; Cttnbp2; CTTNBP2 intron retention form; Kiaa1758; mKIAA1758; ORF4 | |
Rabbit | |
Protein A | |
RUO | |
83992 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8WZ74 | |
CTTNBP2 | |
KLH conjugated synthetic peptide derived from human CTTNBP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction