Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CTU1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24750025UL
Description
CTU1 Polyclonal specifically detects CTU1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CTU1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
ATP binding domain 3, ATPBD3cancer-associated protein, Cancer-associated gene protein, cytoplasmic tRNA 2-thiolation protein 1, Cytoplasmic tRNA adenylyltransferase 1, cytosolic thiouridylase subunit 1 homolog (S. pombe), EC 2.7.7, EC 2.7.7.-, MGC17332, NCS6ATP-binding domain-containing protein 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
90353 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CTU1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RLGISLQLVAVDEGIGGYRDAALAAVRRQAARWELPLTVVAYEDLFGGWTMDAVARSTAGSGRSRSCCTFCGVLR | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction