Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599306 Supplier Invitrogen™ Supplier No. PA599306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ATPBD3.

Plays a central role in 2-thiolation of mcm(5)S(2)U at tRNA wobble positions of tRNA(Lys), tRNA(Glu) and tRNA(Gln). Directly binds tRNAs and probably acts by catalyzing adenylation of tRNAs, an intermediate required for 2-thiolation. It is unclear whether it acts as a sulfurtransferase that transfers sulfur from thiocarboxylated URM1 onto the uridine of tRNAs at wobble position.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTU1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CTU1
Gene Accession No. Q7Z7A3
Gene Alias ATP binding domain 3; Atpbd3; ATP-binding domain-containing protein 3; BC005752; Cancer-associated gene protein; cancer-associated protein; CTU1; Cytoplasmic tRNA 2-thiolation protein 1; cytoplasmic tRNA adenylyltransferase 1; cytosolic thiouridylase subunit 1; cytosolic thiouridylase subunit 1 homolog; cytosolic thiouridylase subunit 1 homolog (S. pombe); NCS6
Gene Symbols CTU1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CTU1(Accession Q7Z7A3), corresponding to amino acid residues L298-F348.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90353
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.