Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CTU2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588908 Supplier Invitrogen™ Supplier No. PA588908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-431, Jurkat; Cellular Location: Cytoplasm.

Cytoplasmic tRNA 2-thiolation protein 2 (CUT2), also named as C16orf84 or NCS2, is a 515 amino acid protein, which belongs to the CUT2 family. C16orf84 localizes in the cytoplasm and Plays a central role in 2-thiolation of mcm5S2U at tRNA wobble positions of tRNA(Lys), tRNA(Glu) and tRNA(Gln). C16orf84 may act by forming a heterodimer with CTU1/ATPBD3 that ligates sulfur from thio-carboxylated URM1 onto the uridine of tRNAs at wobble position.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTU2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CTU2
Gene Accession No. Q2VPK5
Gene Alias 2310061F22Rik; C16orf84; Ctu2; Cytoplasmic tRNA 2-thiolation protein 2; cytoplasmic tRNA 2-thiolation protein 2 {ECO:0000255; cytosolic thiouridylase subunit 2; cytosolic thiouridylase subunit 2 homolog; cytosolic thiouridylase subunit 2 homolog (S. pombe); HAMAP-Rule:MF_03054}; Ncs2; UPF0432
Gene Symbols CTU2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 348180
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.