Learn More
Invitrogen™ CU070 Polyclonal Antibody

Description
CU070 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | CU070 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FAM207A |
| Gene Accession No. | P58468, Q9NSI2 |
| Gene Alias | 1810008A18Rik; AI303072; C21orf70; Fam207a; family with sequence similarity 207 member A; family with sequence similarity 207, member A; PRED56; Protein FAM207A |
| Gene Symbols | FAM207A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.