Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genomadix™ Cube CYP2C19 System

Catalog No. 22-040-051
Click to view available options
:
System

The Cube CYP2C19 System is an FDA 510(K)-cleared, sample-to-result DNA testing system with integrated DNA extraction and amplification.

  • FDA 510(k)-cleared.
  • The Genomadix Cube is a thermal cycling instrument that automatically integrates extraction of DNA from the buccal sample, PCR amplification and fluorescence-based detection of CYP2C19 alleles
  • The computer is a consumer-grade netbook computer that serves as the user interface for logging into the platform, inputting patient information, starting the test, and genotype calling.
  • The barcode scanner is used to scan test kit barcodes (used for recording lot and expiry information) and for scanning patient identifier barcodes.
  • No maintenance or calibration required.

Specifications

Clia Complexity High
Type Genotyping System
For Use With (Application) Determine an individual's CYP2C19 genotype in relation to the 2, 3 and 17 loci
Form Ready-to-Use (RTU)

FDA 510(K)-cleared

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.