|
14-561-188
|
1.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|
|
14-561-155
|
320 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-153
|
620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-152
|
1.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-150
|
2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-149
|
3.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-147
|
5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-154
|
150 g, 300 g, 620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-151
|
1.01 kg, 2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-148
|
1.2 kg, 2.4 kg, 5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield A: Motorized
|
|
14-561-164
|
320 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-162
|
620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-161
|
1.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-159
|
2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-158
|
3.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-156
|
5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-163
|
150 g, 300 g, 620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-160
|
1.01 kg, 2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-157
|
1.2 kg, 2.4 kg, 5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield E: Small glass
|
|
14-561-173
|
320 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-171
|
620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-170
|
1.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-168
|
2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-167
|
3.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-165
|
5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-172
|
150 g, 300 g, 620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-166
|
1.2 kg, 2.4 kg, 5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-169
|
1.01 kg, 2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield I: Motorized with ionizer
|
|
14-561-182
|
320 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield R: Stainless steel frame
|
|
14-561-265
|
2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
|
Draft shield R: Stainless steel frame
|
|
14-561-269
|
3.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report, QP99 software
|
Draft shield R: Stainless steel frame
|
|
14-561-174
|
5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield R: Stainless steel frame
|
|
14-561-181
|
150 g, 300 g, 620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield R: Stainless steel frame
|
|
14-561-178
|
1.01 kg, 2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield R: Stainless steel frame
|
|
14-561-175
|
1.2 kg, 2.4 kg, 5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield R: Stainless steel frame
|
|
14-561-190
|
150 g, 300 g, 620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|
|
14-561-191
|
320 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|
|
14-561-189
|
620 g
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|
|
14-561-186
|
2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|
|
14-561-185
|
3.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|
|
14-561-183
|
5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|
|
14-561-187
|
1.01 kg, 2.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|
|
14-561-184
|
1.2 kg, 2.4 kg, 5.2 kg
|
Balance, Accessories, Power supply, Manual, Factory test report
|
Draft shield U: Manual glass
|