Learn More
Sartorius Cubis™ II QApps
Catalog No. 14561218 Shop All Sartorius Products
Change view
Click to view available options
Type:
Connectivity Package QP4
Pharma Package QP1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-561-218 Pharma Package QP1
14-561-221 Connectivity Package QP4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-561-218 Supplier Sartorius Supplier No. QP1
QApp-Packages are software programs (QApps), for instance, lab applications, functions, or utilities.
  • After licensing, QApps can be activated and run in a task
  • Licenses for the QApp-Packages can be ordered together with the balance
  • All packages are pre-installed on the balance and ready for use after delivery
  • In the QApp-Center, browse through the packages to get detailed information about content
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cubis Balances
For Use With (Application) User management, digital signatures, audit trail, USP minimum weight
Includes Applications concerning the topic compliance with pharmaceuticalrelevant guidelines as 21 CFR Part 11 and USP 39, Chapter 41.
Product Line QApp-Packages
Type Pharma Package QP1