Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUBN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567616 Supplier Invitrogen™ Supplier No. PA567616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CUBN Polyclonal Antibody for Western Blot, IHC (P)

Cubilin (CUBN) acts as a receptor for intrinsic factor-vitamin B12 complexes. The role of receptor is supported by the presence of 27 CUB domains. Cubulin is located within the epithelium of intestine and kidney. Mutations in CUBN may play a role in autosomal recessive megaloblastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUBN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CUBN
Gene Accession No. O60494
Gene Alias 460 kDa receptor; AA408369; AL022750; cubilin; cubilin (intrinsic factor-cobalamin receptor); cubilin precursor variant 1; cubilin precursor variant 2; cubilin precursor variant 3; CUBN; Cubulin; D2Wsu88e; Glycoprotein 280; GP280; IFCR; Intestinal intrinsic factor receptor; intrinsic factor-cobalamin receptor; Intrinsic factor-vitamin B12 receptor; MGA1
Gene Symbols CUBN
Host Species Rabbit
Immunogen Synthesized peptide derived from human CUBN.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8029
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.