Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CUBN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583684
Description
Immunogen sequence: SDQGFHITYL TSPSDLRCGG NYTDPEGELF LPELSGPFTH TRQCVYMMKQ PQGEQIQINF THVELQCQSD SSQNYIEVRD GETLLGKVCG NGTISHIKSI TNSVWIRFKI DASVEKASFR AVYQVACGDE LTGEGVIRSP FFPNVYPGE.
Cubilin (CUBN) acts as a receptor for intrinsic factor-vitamin B12 complexes. The role of receptor is supported by the presence of 27 CUB domains. Cubulin is located within the epithelium of intestine and kidney. Mutations in CUBN may play a role in autosomal recessive megaloblastic anemia.
Specifications
CUBN | |
Polyclonal | |
Unconjugated | |
CUBN | |
460 kDa receptor; AA408369; AL022750; cubilin; cubilin (intrinsic factor-cobalamin receptor); cubilin precursor variant 1; cubilin precursor variant 2; cubilin precursor variant 3; CUBN; Cubulin; D2Wsu88e; Glycoprotein 280; GP280; IFCR; Intestinal intrinsic factor receptor; intrinsic factor-cobalamin receptor; Intrinsic factor-vitamin B12 receptor; MGA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8029 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60494 | |
CUBN | |
Recombinant protein corresponding to Human CUBN. Recombinant protein control fragment (Product # a href=. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction