Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUBN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547527
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547527 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547527 Supplier Invitrogen™ Supplier No. PA547527
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Notch-2 and rhNotch-3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Cubilin (CUBN) acts as a receptor for intrinsic factor-vitamin B12 complexes. The role of receptor is supported by the presence of 27 CUB domains. Cubulin is located within the epithelium of intestine and kidney. Mutations in CUBN may play a role in autosomal recessive megaloblastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUBN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CUBN
Gene Accession No. O60494, O70244, Q9JLB4
Gene Alias 460 kDa receptor; AA408369; AL022750; cubilin; cubilin (intrinsic factor-cobalamin receptor); cubilin precursor variant 1; cubilin precursor variant 2; cubilin precursor variant 3; CUBN; Cubulin; D2Wsu88e; Glycoprotein 280; GP280; IFCR; Intestinal intrinsic factor receptor; intrinsic factor-cobalamin receptor; Intrinsic factor-vitamin B12 receptor; MGA1
Gene Symbols CUBN
Host Species Sheep
Immunogen E. coli-derived recombinant human Cubilin Ser36-Gly126.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65969, 8029, 80848
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.