Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cucurbitacin I
GREENER_CHOICESDP

Catalog No. 15711 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-711 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 15-711 Supplier Tocris Bioscience™ Supplier No. 1571/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of STAT3/JAK2 signaling

Selective inhibitor of STAT3/JAK2 signaling. Inhibits the activation of STAT3 and JAK2 and displays no activity on Src, Akt, ERK and JNK. Suppresses phosphotyrosine levels of STAT3, inhibits STAT3 DNA binding and STAT3-mediated gene expression. Induces apoptosis in cell lines expressing constitutively active tyrosine-phosphorylated STAT3.

Chemical Identifiers

CAS 2222-07-3
Molecular Formula C30H42O7

Specifications

Quantity 1 mg
Formula Weight 514.65
Purity >98%
Molecular Weight (g/mol) 514.66
Chemical Name or Material Cucurbitacin I
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.