Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUEDC2 Antibody (003), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289902PCP
Description
CUEDC2 Monoclonal antibody specifically detects CUEDC2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
CUEDC2 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
003 | |
ELISA, Immunohistochemistry-Paraffin | |
bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CUEDC2. | |
0.1 mL | |
Signal Transduction | |
79004 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction