Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUEDC2 Antibody (08), Alexa Fluor™ 647 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306373AF647
Description
CUEDC2 Monoclonal antibody specifically detects CUEDC2 in Human samples. It is validated for Flow CytometrySpecifications
CUEDC2 | |
Monoclonal | |
Alexa Fluor 647 | |
bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CUEDC2 (rh CUEDC2; NP_076945.2; aa1-287). | |
0.1 mL | |
Signal Transduction | |
79004 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
08 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction