Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUEDC2 Rabbit anti-Human, Clone: 3, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | CUEDC2 |
---|---|
Clone | 3 |
Dilution | ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000 |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
CUEDC2 Monoclonal specifically detects CUEDC2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CUEDC2 | |
ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
79004 | |
IgG | |
Protein A purified |
3 | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
Recombinant Human CUEDC2 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title