Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CUG-BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14130
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14130 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14130 Supplier Invitrogen™ Supplier No. PA14130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4130 detects ETR-3 in human and mouse samples. PA1-4130 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~50 kDa protein representing ETR-3 in heart lysate. The PA1-4130 immunogen is a synthetic peptide corresponding to the C-terminal residues V(478) Q L K R S K N D S K P Y C(490) of human ETR3 (a.a. 496-508 in mouse).

CUG-BP1 and CUG-BP2 are proteins that bind specifically to (CUG)8 oligonucleotides in vitro. While CUG-BP1 has the major binding activity in normal cells, nuclear CUG-BP2 binding activity increases in DM cells. Both CUG-BP1 and CUG-BP2 are isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. CUG-BP1, an RNA CUG triplet repeat binding protein, regulates splicing and translation of various RNAs. Expansion of RNA CUG repeats in the DMPK in DM is associated with alterations in binding activity of CUG-BP1 as well as alterations in the translation of the C/EBPb transcription factor. CUG-BP1 is an important regulator of initiation from different AUG codons of C/EBPb mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CUG-BP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CELF2
Gene Accession No. O95319, Q9Z0H4
Gene Alias B230218O03; B230345P09Rik; BRUNOL3; Bruno-like protein 3; C88023; cb906; CELF 2; Celf2; CELF-2; CUG triplet repeat RNA-binding protein 2; CUG triplet repeat, RNA binding protein 2; CUG-BP- and ETR-3-like factor 2; Cugbp- and Etr3-like factor 2; CUGBP Elav-like family member 2; CUGBP, Elav-like family member 2; Cugbp2; CUG-BP2; cugbp2 protein; D230046B21Rik; elav-type ribonucleoprotein-3; elav-type RNA-binding protein 3; ETR3; ETR-3; fj87b07; hNAPOR; mETR-3; mNapor; NAPOR; napor protein isoform; napor protein long isoform; napor protein short isoform; Napor-2; neur; neuroblastoma apoptosis-related RNA-binding protein; RNA-binding protein; RNA-binding protein Brunol3; RNA-binding protein BRUNOL-3; rNapor; RP23-124P7.1; wu:fj87b07
Gene Symbols CELF2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal residues V(478) Q L K R S K N D S K P Y C(490) of human ETR3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10659, 14007
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.