Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUGBP1/CELF1 Antibody (3B1), Novus Biologicals™
SDP

Catalog No. p-4974352 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200316SS 0.025 mL
NB200316 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB200316SS Supplier Novus Biologicals Supplier No. NB200316SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 11 publications

CUGBP1/CELF1 Monoclonal specifically detects CUGBP1/CELF1 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays.

Specifications

Antigen CUGBP1/CELF1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3B1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Simple Western 1:200, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, Gel Super Shift Assays reported in scientific literature
Gene Accession No. Q92879
Gene Alias BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2
Gene Symbols CELF1
Host Species Mouse
Immunogen Full-length human CUGBP1 [UniProt# Q92879]
Molecular Weight of Antigen 50 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Neurodegeneration
Primary or Secondary Primary
Gene ID (Entrez) 10658
Target Species Human, Mouse, Rat, Pig, Bovine, Primate, Rabbit
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.