Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUGBP1/CELF1 Antibody (OTI5B8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271623H
Description
CUGBP1/CELF1 Monoclonal specifically detects CUGBP1/CELF1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CUGBP1/CELF1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CELF1 | |
Full length recombinant protein of human CUGBP1/CELF1 (NP_006551) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI5B8 | |
Western Blot, Immunohistochemistry | |
BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2 | |
Mouse | |
Affinity Purified | |
Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Neurodegeneration, Phospho Specific | |
10658.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction