Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUGBP1/CELF1 Antibody (3B1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280684
Description
CUGBP1/CELF1 Monoclonal specifically detects CUGBP1/CELF1 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Gel Super Shift Assays, Immunocytochemistry.Specifications
CUGBP1/CELF1 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1:500, Simple Western : 1:200, Flow Cytometry : 1 ug per million cells, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence : 1:50-1:200, Immunoprecipitation reported in scientific literature, Immunohistochemistry-Paraffin : 1:100-1:500, Immunohistochemistry-Frozen : 1:100-1:500, Gel Super Shift Assays reported in scientific literature, Immunocytochemistry | |
BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Primate, Rabbit | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Gel Shift | |
3B1 | |
Unconjugated | |
Tris-Glycine, 0.15 M NaCl | |
CELF1 | |
Full-length human CUGBP1 [UniProt# Q92879] | |
0.1 mL | |
Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Neurodegeneration, Phospho Specific | |
10658 | |
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction