Learn More
CUGBP1 Monoclonal Antibody (OTI5B8)

Description
CUGBP1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | CUGBP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI5B8 |
| Concentration | 0.99 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CELF1 |
| Gene Accession No. | Q92879 |
| Gene Alias | 1600010O03Rik; 50 kDa nuclear polyadenylated RNA-binding protein; AA407467; Brain protein F41; Brunol2; bruno-like 2; bruno-like protein 2; Celf1; CELF-1; CUG RNA-binding protein; CUG triplet repeat RNA-binding protein 1; CUG triplet repeat, RNA binding protein 1; CUG triplet repeat, RNA-binding protein 1; Cugbp; CUG-BP; CUG-BP- and ETR-3-like factor 1; CUGBP Ela; CUGBP Elav-like family member 1; CUGBP, Elav-like family member 1; CUGBP, Elav-like family member 1-like; CUGBP1; CUG-BP1; D2Wsu101e; Deadenylation factor CUG-BP; deadenylation factor EDEN-BP; EDEN-BP; EDEN-BP homolog; embryo deadenylation element binding protein; embryo deadenylation element-binding protein homolog; HNAB50; NAB50; NAPOR; nuclear polyadenylated RNA-binding protein, 50-kD; RNA-binding protein BRUNOL-2; RP23-147D3.7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.