Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUGBP2 Mouse anti-Human, Clone: PCRP-CELF2-1E4, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | CUGBP2 |
---|---|
Clone | PCRP-CELF2-1E4 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array |
Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
CUGBP2 Monoclonal antibody specifically detects CUGBP2 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein ArraySpecifications
CUGBP2 | |
0.2 mg/ml | |
Flow Cytometry, Immunofluorescence, Peptide Array | |
Unconjugated | |
Mouse | |
Lipid and Metabolism, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
BRUNOL3ELAV-type RNA-binding protein 3, Bruno-like protein 3, CELF-2, CUG triplet repeat RNA-binding protein 2, CUG triplet repeat, RNA binding protein 2, CUG triplet repeat, RNA-binding protein 2, CUG-BP- and ETR-3-like factor 2, CUGBP Elav-like family member 2, CUGBP, Elav-like family member 2, CUG-BP2, CUGBP2hNAPOR, Etr-3, ETR3RNA-binding protein BRUNOL-3, NAPOR, NAPOR-2, Neuroblastoma apoptosis-related RNA-binding protein | |
Recombinant full-length human CUGBP2 protein (Uniprot: O95319) | |
Primary | |
Store at 4°C. |
PCRP-CELF2-1E4 | |
Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
O95319 | |
10659 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title