Learn More
Cul1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Cul1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-4969
![]() |
Bethyl Laboratories, Inc
A303373AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Cullin 1 is a member of the family of human cullin genes (CUL 1, 2, 3, 4a, 4b, and 5) homologous to the S. cerevisiae cdc 53 gene. CUL 1 forms a complex with human p19 skp1 and F box protein p45 skp2, both of which, together with cdc 53 (known as CUL A), as three subunits form a ubiquitin ligase controlling proteolysis of G1 cell cycle regulatory proteins. The CUL 1/p19 skp1/p45 skp2complex is thought to play a role in the ubiquitination of G1 regulatory proteins.Specifications
Cul1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CUL-1 | |
Between 726 and 776 | |
Primary | |
4° C | |
CUL1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q13616, Q9WTX6 | |
26965, 8454 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.