Learn More
Cul2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$137.00
Specifications
| Antigen | Cul2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2687
|
Bethyl Laboratories, Inc
A302476AT |
20 μL |
Each of 1 for $137.00
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan
CUL-2 is a member of the family of human cullin genes (CUL-1, -2, -3, -4a, -4b, and -5), homologous to the S. cerevisiae cdc53 gene. CUL-1 forms a complex with human p19skp1 and F box protein p45skp2. Together with cdc53 (Known as CulA), as three subunits, they form a ubiquitin ligase controlling proteolysis of G1 cell cycle regulatory proteins. In contrast to CUL-1, CUL-2 does not associate with p19skp1 and p45skp2. Instead, CUL-2 forms a complex with an inactive transcriptional elongation complex, SIII, formed by three subunits: Elongain C, Elongin B and VHL. The function of this association remains unknown.Specifications
| Cul2 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| CUL-2 | |
| CUL2 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13617 | |
| 8453 | |
| Between 695 and 745 | |
| Primary | |
| 4° C | |
| CUL2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.