Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cul2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562685
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2685 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2685 Supplier Bethyl Laboratories, Inc Supplier No. A302476A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan

CUL-2 is a member of the family of human cullin genes (CUL-1, -2, -3, -4a, -4b, and -5), homologous to the S. cerevisiae cdc53 gene. CUL-1 forms a complex with human p19skp1 and F box protein p45skp2. Together with cdc53 (Known as CulA), as three subunits, they form a ubiquitin ligase controlling proteolysis of G1 cell cycle regulatory proteins. In contrast to CUL-1, CUL-2 does not associate with p19skp1 and p45skp2. Instead, CUL-2 forms a complex with an inactive transcriptional elongation complex, SIII, formed by three subunits: Elongain C, Elongin B and VHL. The function of this association remains unknown.

Specifications

Antigen Cul2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene CUL2
Gene Accession No. Q13617
Gene Alias CUL-2
Gene Symbols CUL2
Host Species Rabbit
Immunogen Between 695 and 745
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8453
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.