Learn More
CUL4A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | CUL4A |
---|---|
Concentration | 0.3 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9777
![]() ![]() |
Proteintech Group Inc
148511AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specifically against CUL4A,but not CUL4B. CUL4A has 2 isoforms with MW 88 kDa (759 aa) and 77 kDa (659 aa), and 14851-1-AP recognizes both.
CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.Specifications
CUL4A | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q13619, Q3TCH7 | |
361181, 8451, 99375 | |
Peptide | |
Primary | |
-20°C | |
CUL4A |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CUL 4A, CUL4A, cullin 4A | |
Cul4a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.