Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUL4B Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297399AF647
Description
CUL4B Polyclonal specifically detects CUL4B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CUL4B | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
CUL-4B, cullin 4B, cullin-4B, DKFZp686F1470, KIAA0695, MRXHF2, MRXS15, MRXSC, SFM2 | |
Produced in rabbits immunized with E. coli-derived Human CUL4B fragment. | |
0.1 mL | |
Cell Cycle and Replication | |
8450 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction