Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUL4B Antibody (OTI1C4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271373AF750
Description
CUL4B Monoclonal specifically detects CUL4B in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CUL4B | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CUL4B | |
Recombinant protein fragment corresponding to amino acids 1-300 of human CUL4B (NP_003579) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1C4 | |
Western Blot, Immunohistochemistry | |
CUL-4B, cullin 4B, cullin-4B, DKFZp686F1470, KIAA0695, MRXHF2, MRXS15, MRXSC, SFM2 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
8450.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction