Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cullin 1 Antibody (AS97.1), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002616JF646
Description
Cullin 1 Monoclonal antibody specifically detects Cullin 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
Cullin 1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
AS97.1 | |
Western Blot | |
CUL-1, cullin 1, cullin-1, MGC149834, MGC149835 | |
Bacterially expressed full length recombinant human Cullin 1 | |
0.1 mL | |
Apoptosis, Cell Cycle and Replication | |
8454 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction