Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cullin 1 Recombinant Rabbit Monoclonal Antibody (5I5Z3)
GREENER_CHOICE

Catalog No. PIMA535216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535216 Supplier Invitrogen™ Supplier No. MA535216
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: NKKLRVNINV PMKTEQKQEQ ETTHKNIEED RKLLIQAAIV RIMKMRKVLK HQQLLGEVLT QLSSRFKPRV PVIKKCIDIL IEKEYLERVD GEKDTYSYLA.

CUL1 gene ontology annotations related to this gene include animal organ morphogenesis; apoptotic process; cell proliferation; protein monoubiquitination; SCF-dependent proteasomal ubiquitin-dependent protein catabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 5I5Z3
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CUL1
Gene Accession No. Q13616, Q9WTX6
Gene Alias Cul1; CUL-1; cullin 1; cullin-1
Gene Symbols CUL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 677-776 of human Cullin 1 (Q13616).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26965, 362356, 8454
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.