Learn More
Invitrogen™ Cullin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120792
Description
Positive test controls include: A-549, MCF7, NIH/3T3, K-562, C6, Mouse liver, Mouse brain, Rat testis. Immunogen sequence: KNPEEAELED TLNQVMVVFK YIEDKDVFQK FYAKMLAKRL VHQNSASDDA EASMISKLKQ ACGFEYTSKL QRMFQDIGVS KDLNEQFKKH LTNSEPLDLD FSIQVLSSGS WPFQQSCTFA LPSELERSYQ RFTAFYASRH SGRKLTWLYQ LSKGELVTN.
CUL1 gene ontology annotations related to this gene include animal organ morphogenesis; apoptotic process; cell proliferation; protein monoubiquitination; SCF-dependent proteasomal ubiquitin-dependent protein catabolic process.
Specifications
Cullin 1 | |
Polyclonal | |
Unconjugated | |
CUL1 | |
Cul1; CUL-1; cullin 1; cullin-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26965, 362356, 8454 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.88 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13616, Q9WTX6 | |
CUL1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 435-593 of human Cullin 1 (NP_0035832). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.