Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cullin 3, Rabbit anti-Human,Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP280431
Description
Cullin 3 Polyclonal specifically detects Cullin 3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cullin 3 | |
Polyclonal | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide | |
CUL3 | |
Synthetic peptide within human Cullin 3 aa 724-768. (SwissProt: Q13618 Human; SwissProt: Q9JLV5 Mouse) | |
100 μL | |
Apoptosis, Cell Cycle and Replication | |
8452 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200Flow 1:50-1:100 | |
CUL-3, cullin 3, cullin-3, KIAA0617 | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction